Merck
Z117048
Stainless steel 304 syringe needle
gauge 14, L 10 in.
About This Item
Código UNSPSC:
41122002
eCl@ss:
32070500
NACRES:
NB.03
descripción
Standard Hub with 304 Stainless Steel Tubing
envase
pack of 1 ea
fabricante / nombre comercial
Cadence Science
Longitud
10 in.
manómetro
14
Descripción general
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)