Key Documents
Z103314
Sigma-Aldrich
Aldrich® round-bottom flask with septum-inlet
capacity 100 mL, joint: ST/NS 14/20
Sinónimos:
bottom flask, flask round, round flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
round bottom flask
two-neck
joint
joint: ST/NS14/20
sterility
non-sterile
feature
closure type Standard Taper joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
100 mL
General description
Mit NS Schliff. Passende Septen: Z10,072-2 oder Z12,435-4 (siehe Abschnitt Septen).
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)