Key Documents
WHA10350220
Whatman® cellulose extraction thimbles
603 Cellulose thimbles, 26 x 60 mm - thickness 1.5 mm, 25/pk
Sinónimos:
Z696420, cellulose thimble, extraction thimble, glass thimbles
About This Item
UNSPSC Code:
41104817
NACRES:
NB.43
packaging
pack of 25
manufacturer/tradename
Whatman 10350220
Whatman Article No. 28414687 (US reference)
W × L
26 mm × 60 mm
thickness
1.5 mm
Features and Benefits
  • 603 Standard Cellulose Extraction Thimbles made from high-quality cellulose available in a range of sizes.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)