The pressure rating for this item is 88psi.
SYNP160001
Synthware™ cylindrical pressure vessel, heavy wall
15 mL, O.D. × L 26 mm × 109 mm, #15 bushing
Sinónimos:
Heavy-Wall Pressure Vessel, Synthware P160001, pressure tube
About This Item
Productos recomendados
envase
pkg of 1 ea
fabricante / nombre comercial
Synthware P160001
D.E. × L
26 mm × 109 mm
capacidad
15 mL
Adaptador
#15 bushing
Información legal
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Los clientes también vieron
-
what is the pressure rating of these bottles
1 answer-
Helpful?
-
-
What size of Viton O-ring is suitable for Synthware™ cylindrical pressure vessel, heavy wall, 15 ml (product number SYNP160001-1EA)?
1 answer-
Unfortunately, replacement O-rings for this product are not available at this time. These items are available directly from the manufacturer under Synthware product number R111EP. Please see the link below to the supplier product page:
https://kemtech-america.com/Product/Product_Info.aspx?ProductID=3033Helpful?