Documentos clave
SYNF904200G
Synthware pear-shaped Schlenk flask with stopcock
capacity 200 mL, joint: ST/NS 24/40
Sinónimos:
Synthware F904200G, modified Schlenk reaction flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
material
borosilicate glass flask
pear-shaped
single-neck
joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F904200G
capacity
200 mL
General description
A suffix of G indicates a glass stopcock, H indicates a high-vacuum valve, no letter suffix indicates a PTFE Stopcock.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)