Key Documents
PL800GH-9-6
Plas-Labs Hypalon glove box gloves
for 1 piece glove set, size 9 (for 6 in. ports)
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
material
Hypalon®
size
9 (for 6 in. ports)
feature
reusable
packaging
pack of 1 pair
manufacturer/tradename
Plas-Labs 800GH-9-6
General description
Plas-Labs white Hypalon® rubber for resistance to abrasion, oxidizing chemicals, ozone and UV light
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Legal Information
HaackerCalc is a trademark of ChemSW, Inc.
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)