Documentos clave
DWK73662-25
KIMBLE® KIM-KAP® push cap
red polypropylene cap, cap diam. (25 mm)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
red polypropylene cap
sterility
non-sterile
packaging
case of 500 ea
manufacturer/tradename
Kimble® 73662-25
cap diam.
(25 mm)
General description
  • KIM-KAP polypropylene closures for culture tubes
  • Autoclavable
  • Color coded
Features and Benefits
• Autoclavable• Color coded
Legal Information
KIM-KAP is a registered trademark of DWK LIFE SCIENCES LLC
KIMBLE is a registered trademark of DWK Life Sciences
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)