Key Documents
CLS2480400
Pyrex® Allihn condenser, with male and female joints
jacket L 400 mm, joint: ST/NS 24/40
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
joint
joint: ST/NS24/40
manufacturer/tradename
Corning 2480-400
jacket L
400 mm
General description
Suitable for use in vacuum distillation. With drip-tip
Legal Information
PYREX is a registered trademark of Corning, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)