Key Documents
CLS1261CO
Pyrex® 200mL heavy wall centrifuge bottle with GP 38-400 screw cap
white polypropylene replacement caps for 200mL heavy wall centrifuge bottles (CLS1261200)
About This Item
UNSPSC Code:
41103913
NACRES:
NB.43
material
polypropylene cap
white polypropylene
sterility
non-sterile
feature
cap
packaging
case of 1
manufacturer/tradename
Corning 1261-CO
fitting
38-400 PTFE liner
General description
Note: These polypropylene GPI 38-400 screw caps are replacement caps for the 200mL heavy wall centrifuge bottles (CLS1261200) and have PTFE liners.
Particularly useful in handling sputum specimens. The bottle enables digestion, shaking, neutralizing and centrifuging to be performed in one container. The tight-fitting polypropylene screw cap with PTFE liner reduces the possibility of aerosol escape.
Legal Information
PYREX is a registered trademark of Corning, Inc.
For use with
Referencia del producto
Descripción
Precios
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)