Key Documents
CLS12203X
Pyrex® aspirator bottle with outlet for tubing
capacity 13250 mL
Sinónimos:
glass bottle
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
clear bottle
cylindrical bottle
wide-mouth bottle
feature
autoclavable
cap: no
manufacturer/tradename
Corning 1220-3X
capacity
13250 mL
diam. × H
238 mm × 445 mm
Legal Information
PYREX is a registered trademark of Corning, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)