Documentos clave
BR781535
Brand®1.2 ML Tubes
strip of 8 caps, polyethylene (cap)
Sinónimos:
Accessories, Caps, Sample Storage, Tube racks, Tubes
About This Item
Código UNSPSC:
41122804
NACRES:
NB.22
Materiales
polyethylene (cap)
polypropylene (tubes and rack)
esterilidad
non-sterile
Características
strip of 8 caps
envase
pack of 120
fabricante / nombre comercial
BRAND 781535
capacidad
1.2 mL
Descripción general
BIO-CERT® CERTIFIED QUALITY: Free of human DNA, RNase, DNase and Pyrogens
Información legal
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)