Documentos clave
BR4600310
BRAND® Dispensette® S Digital bottle-top dispenser
volume range 0.1-1 mL, without recirculation valve
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
Características
incremental 0.005 mL
DE-M marking
without recirculation valve
fabricante / nombre comercial
BRAND 4600310
intervalo de volumen
0.1-1 mL
Adaptador
(GL 28/S28)
GL 32
GL 38
S 40
Descripción general
Dispensette S is suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents.
Características y beneficios
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Otras notas
Digital-The mechanical volume setting is easy to read and ensures accurate and reproducible volume control. Easy Calibration Technique for calibration adjustment in seconds without tools. A calibration flag indicates changes from factory settings.
Información legal
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Protocolos
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Contenido relacionado
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.