Merck
  • High-level fed-batch fermentative expression of an engineered Staphylococcal protein A based ligand in E. coli: purification and characterization.
High-level fed-batch fermentative expression of an engineered Staphylococcal protein A based ligand in E. coli: purification and characterization.
AMB Express (2015-11-12)
Martin Kangwa, Vikas Yelemane, Ayse Nur Polat, Kanaka Durga Devi Gorrepati, Mariano Grasselli, Marcelo Fernández-Lahore
ABSTRACT
The major platform for high level recombinant protein production is based on genetically modified microorganisms like Escherichia coli (E. coli) due to its short dividing time, ability to use inexpensive substrates and additionally, its genetics is comparatively simple, well characterized and can be manipulated easily. Here, we investigated the possibilities of finding the best media for high cell density fermentation, by analyzing different media samples, focusing on improving fermentation techniques and recombinant protein production. Initial fermentation of E. coli BL21 DE3:pAV01 in baffled flasks showed that high cell density was achieved when using complex media, Luria-Bertani (LB) and Terrific medium broth (TB) (10 and 14 g/L wet weight, respectively), as compared to mineral media M9, modified minimal medium (MMM) and Riesenberg mineral medium (RM) (7, 8 and 7 g/L, respectively). However, in fed-batch fermentation processes when using MMM after 25 h cultivation, it was possible to yield an optical density (OD600) of 139 corresponding to 172 g/L of wet biomass was produced in a 30 L TV Techfors-S Infors HT fermenter, with a computer controlled nutrient supply (glucose as a carbon source) delivery system, indicating nearly 1.5 times that obtained from TB. Upon purification, a total of 1.65 mg/g of protein per gram cell biomass was obtained and the purified AviPure showed affinity for immunoglobulin. High cell density fed batch fermentation was achieved by selecting the best media and growth conditions, by utilizing a number of fermentation parameters like media, fermentation conditions, chemical concentrations, pO2 level, stirrer speed, pH level and feed media addition. It is possible to reach cell densities higher than shake flasks and stirred tank reactors with the improved oxygen transfer rate and feed.
Product Description
Supelco
Terrific Broth, modified, powder