Skip to Content
Merck
All Photos(1)

Key Documents

G005854

LC/MS Analysis of Antiarrhythmic Drugs and Metabolites in Plasma on Ascentis® Express HILIC following Sample Prep using HybridSPE®-Phospholipid

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

sample preparation: SPE (Solid Phase Extraction)
sample/matrix: rabbit plasma, unfiltered K2-EDTA, spiked with each compound at 100 ng/mL (3:1, plasma:1% formic acid in acetonitrile)
SPE tube/cartridge: HybridSPE-Phospholipid, 96-well plate (575656-U)
sample addition: to each well add 100 μL plasma, followed by a 300 μL of 1% formic acid in acetonitrile
elution: collect, filtrate and analyze directly.
column: Ascentis Express HILIC, 10 cm x 2.1 mm I.D., 2.7 μm particles (53939-U)
mobile phase: [A] 5 mM ammonium formate; [B] 5 mM ammonium formate in acetonitrile, 5:95 (A:B), adjusted to pH 7.0 with formic acid
flow rate: 0.4 mL/min
pressure: 1305 psi (90 bar)
column temp.: 35 °C
detector: ESI+, 100-1000 m/z
injection: 0.5 μL

suitability

application for HPLC

application(s)

clinical

Analysis Note

This application shows the extraction and analysis of antiarrhythmic cardiac drugs in plasma samples using LC/MS/MS. HILIC chromatography on Ascentis Express along with a selective sample preparation using HybridSPE-Phospholipid provided sensitivity and resolution. The highest grade LC-MS solvents were used to supply low background interference and low particulate contaminants for robust, trouble-free operation. Cerilliant CRMs provided reliable identification and quantification.

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany
HybridSPE is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Lidocaine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    Amiodarone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    N-Desethylamiodarone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    (±)-Flecainide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service