Skip to Content
Merck
All Photos(1)

Documents

97729-U

Supelco

Flash Silica Inorganic Sorbent

spherical, 200-400 mesh, pkg of 1 kg, dried

Synonym(s):

Silica gel

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
NC.05

product name

Flash Silica, dried, pkg of 1 kg, spherical

grade

high-purity grade

form

powder

packaging

pkg of 1 kg

manufacturer/tradename

Sigma-Aldrich

technique(s)

LPLC: suitable

surface area

480 m2/g

matrix

Silica

matrix active group

Silica

particle size

200-400 mesh
40-75 μm

pore size

0.7-0.9 cm3/g pore volume
70 Å pore size

bp

2230 °C

mp

>1600 °C

separation technique

hydrophilic interaction (HILIC)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service