Skip to Content
Merck
All Photos(1)

Documents

818008

TSKgel® SWXL (7 µm) HPLC Columns

L x I.D. 4 cm × 6 mm, diol phase

Synonym(s):

TSKgel® BioAssist® SWXL Guard Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

product name

TSKgel® SWXL Columns, phase diol, L × I.D. 4 cm × 6 mm, 7 μm particle size, PEEK hardware

material

PEEK hardware

Agency

suitable for USP L20
suitable for USP L59

product line

TSKgel®

manufacturer/tradename

Tosoh 18008

parameter

≤0.5 M salt concentration
0-100% aqueous soluble organic solvents
1.2 mL/min flow rate
10-30 °C temperature

technique(s)

HPLC: suitable

L × I.D.

4 cm × 6 mm

matrix

silica particle platform

matrix active group

diol phase

particle size

7 μm

pore size

25 Å

operating pH

2.5-7.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel SWXL-type columns contain smaller particles than TSKgel SW-type columns; 5 and 8μm versus 10 and 13μm. As the TSKgel SW-type columns, TSKgel SWXL columns feature highly porous silica particles, the surface of which has been shielded from interacting with proteins by derivatization with ligands containing diol functional groups. TSKgel SWXL-type columns stand out from other silica- or polymer-based high performance size exclusion columns by virtue of their large pore volumes. These columns are commonly used in the quality control of monoclonal antibodies and other biopharma products.

Physical form

Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M phosphate buffer, pH 6.7.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

BioAssist is a registered trademark of Tosoh Corporation
TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service