Skip to Content
Merck
All Photos(1)

Key Documents

58639

Upchurch Fittings Kit

for use with Waters

Synonym(s):

UPTIGHT FITTINGS KIT, WATERS SYSTEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

manufacturer/tradename

Upchurch Scientific, Inc 1413

compatibility

for use with Waters

Looking for similar products? Visit Product Comparison Guide

General description

When caught without the fitting needed to install a column, new plumbing line, etc., these kits come in handy. They contain all the fittings and tubings needed to operate the high performance liquid chromatography (HPLC) system, conveniently stored in a plastic case.

Features and Benefits

; S.S. Waters ZDV Union, 0.020", 2 each ; Waters Male Nut, 10 each ; Waters Ferrule, 10 each ; S.S. ZDV Union, 0.020", 2 each ; S.S. Male Nut, 5 each ; S.S. Female Nut, 5 each ; Tefzel Cap, 2 each ; Delrin Plug, 2 each ; Fingertight II Male Nut (Delrin), 2 each ; PEEK Replacement Ferrule, 4 each ; 1/16 in. Flangeless Male Nut, Black, Delrin, 6 each ; 1/16 in. Flangeless Ferrule, Tefzel, 6 each ; Stainless Steel Tubing, 5cm × 0.01" I.D., 4 each ; Stainless Steel Tubing, 20cm × 0.01" I.D., 4 each ; Stainless Steel Tubing, 5cm × 0.02" I.D., 4 each ; Stainless Steel Tubing, 10cm × 0.02" I.D., 4 each ; Stainless Steel Tubing, 20cm × 0.02" I.D., 4 each ; Stainless Steel Tubing, 10cm × 0.01" I.D., 4 each

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Tee bore 0.020 in., 10-32 thread

57661

Tee

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service