Skip to Content
Merck
All Photos(21)

Key Documents

ZRB1600

Sigma-Aldrich

Anti-PUMA Antibody, clone 1L16 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Bcl-2-binding component 3;isoforms 1/2;JFY-1;p53 up-regulated modulator of apoptosis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1L16, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 20.53 kDa
observed mol wt ~18 kDa

purified by

using Protein A

species reactivity

mouse, human, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1L16 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Bcl-2-binding component 3 (PUMA). It targets an epitope within 17 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the C-terminal region of human Bcl-2-binding component 3 (PUMA).

Application

Quality Control Testing

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PUMA in HepG2 cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PUMA in NIH3T3 and L6 cell lysates.

Affinity Binding Assay: A representative lot of this antibody bound PUMA peptide with a KD of 1.0 x 10-12 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected PUMA in human small intestine tissue sections.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected PUMA in A431, HeLa, HepG2, and NIH3T3 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Bcl-2-binding component 3, isoforms 1/2 (UniProt: Q9BXH1; also known as JFY-1, p53 up-regulated modulator of apoptosis. PUMA) is encoded by the BBC3 (also known as PUMA) gene (Gene ID: 27113) in human. PUMA is a member of the BH3-only Bcl-2 family that is localized in the mitochondria and serves as a critical mediator of p53-dependent and -independent apoptosis induced by a wide variety of stress stimuli and growth factor/cytokine withdrawal. It is normally expressed at a very low level in cells, however, it is rapidly induced under stress conditions. PUMA transduces death signals primarily to the mitochondria, where it acts indirectly on Bax and/or Bak by relieving the inhibition imposed by anti-apoptotic members. It is reported to bind and antagonize all antiapoptotic members of the Bcl-2 family and induce mitochondrial dysfunction, cytochrome c release, and caspase activation. Its BH3 domain, localized to amino acids 137-151, is required for its interaction with Bcl-2 like proteins. The BH3 domains forms an amphipathic a-helical structure that allows it to bind to anti-apoptotic Bcl-2 family proteins. Its C-terminal region contains a hydrophobic domain that directs its mitochondrial localization and both the BH3 domain and mitochondrial localization are shown to be essential induction of apoptosis. PUMA is considered as an essential mediator of p53-dependent and p53-independent apoptosis. Inhibition of PUMA can lead to apoptosis deficiency and increases the risk for cancer development and therapeutic resistance. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Yu, J., and Zhang, L. (2008). Oncogene. 27 (suppl. 1); S71-S83; Yu, J., et al. (2001). Mol Cell. 7(3); 673-682).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service