Z743565
Petri-Seal™ adhesive sealing film
red, L × W 12.5 in. × 32.9 in.
All Photos(1)
About This Item
UNSPSC Code:
41122410
NACRES:
NB.35
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Diversified Biotech PSEAL-108R
L × W
12.5 in. × 32.9 in.
color
red
Related Categories
General description
A sealing tape designed specifically for petri dishes and multiwell plates.
Conforms to the irregular perimeter to seal cover to plate, reduce evaporation, and prevent contamination. Waterproof, chemically resistant, and suitable for continuous use from −40 to +77°C. Heavy 5-mil (0.125 mm) thickness resists abrasion and puncture.
Conforms to the irregular perimeter to seal cover to plate, reduce evaporation, and prevent contamination. Waterproof, chemically resistant, and suitable for continuous use from −40 to +77°C. Heavy 5-mil (0.125 mm) thickness resists abrasion and puncture.
Legal Information
Petri-Seal is a trademark of C.B.S. Scientific Co.