Z720526
Nalgene® diagnostic bottles, style 2035
size 5 mL
Synonym(s):
PETG bottle, PETG bottles, nalgene bottle
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
eCl@ss:
32049200
NACRES:
NB.17
Recommended Products
material
PETG bottle
clear bottle
high-density polyethylene cap
round bottle
wide-mouth bottle
Joint
Joints threaded neck (20-415)
feature
with cap
packaging
pack of 20 ea
manufacturer/tradename
Nalgene® 2035-0005
diam. × H
22 mm × 46 mm
size
5 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Nalgene® diagnostic bottles, style 2035 can be used to perform sterile sampling, storage, and shipment of reagents and buffer solutions. These bottles are made up of non-pyrogenic and non-cytotoxic polyethylene terephthalate copolymer with lined white high-density polyethylene (HDPE ) closures.
Features and Benefits
- Sterile to 10-6 SAL
- Includes a leakproof closure for sample protection.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service