TMO140685
Nunc® Cell-Culture Treated Multidishes
polystyrene, wells 6, without lid, Nunclon® Delta treated, L × W 128 mm × 85 mm, case of 85 ea
Synonym(s):
multi-well plate, treated plate
All Photos(1)
About This Item
UNSPSC Code:
41121812
NACRES:
NB.13
Recommended Products
material
polystyrene
packaging
case of 85 ea
manufacturer/tradename
Nunc 140685
L × W
128 mm × 85 mm
wells
6
Related Categories
General description
Promote maximum adhesion for a broad range of cell types with the Thermo Scientific Nunc Cell-Culture Treated multi-well plates with the Nunclon Delta surface treatment. The open format with the raised well rim enables convenient and secure manipulation of the cells.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)