Skip to Content
Merck
All Photos(1)

Key Documents

SML4045

Sigma-Aldrich

Salvianolic acid A

≥98% (HPLC)

Synonym(s):

(αR)-α-[[(2E)-3-[2-[(1E)-2-(3,4-Dihydroxyphenyl)ethenyl]-3,4-dihydroxyphenyl]-1-oxo-2-propen-1-yl]oxy]-3,4-dihydroxybenzenepropanoic acid, (+)-Salvianolic acid A, Dan phenolic acid A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H22O10
CAS Number:
Molecular Weight:
494.45
MDL number:
UNSPSC Code:
41116107
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O([C@H](Cc3cc(c(cc3)O)O)C(=O)O)C(=O)\C=C\c1c(c(c(cc1)O)O)\C=C\c2cc(c(cc2)O)O

InChI key

YMGFTDKNIWPMGF-UCPJVGPRSA-N

Biochem/physiol Actions

Cell protective antioxidant isolated from Salvia.Salvianolic acid A is cell protective antioxidant originally isolated from Salvia. It has been found to attenuate PDGF-induced ROS formation in hepatic stellate cells and protected against isoproterenol-induced myocardial damage in rats.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fan Tang et al.
Antioxidants (Basel, Switzerland), 12(4) (2023-04-28)
The vast majority of drug-induced liver injury is mainly attributed to acetaminophen (APAP) overdose. Salvianolic acid A (Sal A), a powerful water-soluble compound obtained from Salvia miltiorrhiza, has been confirmed to exert hepatoprotective effects. However, the beneficial effects and the
Ji Zhu et al.
Frontiers in pharmacology, 13, 1009229-1009229 (2022-11-26)
Rescuing endothelial cells from pyroptotic cell death emerges as a potential therapeutic strategy to combat diabetic atherosclerosis. Salvianolic acid A (SAA) is a major water-soluble phenolic acid in the Salvia miltiorrhiza Bunge, which has been used in traditional Chinese medicine
Xiaxia Fan et al.
Journal of ethnopharmacology, 308, 116219-116219 (2023-02-10)
Salvianolic acid A (SAA) is the main active component of the classic anti-atherosclerotic drug Salvia miltiorrhiza Bunge. Inflammation-induced infiltration of monocyte/macrophages into the vascular wall is the initiating step in atherogenesis, and targeted blocking of this step may provide a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service