Skip to Content
Merck
All Photos(1)

Key Documents

SML2915

Sigma-Aldrich

SAR-20347

≥98% (HPLC)

Synonym(s):

2-(2-Chloro-6-fluorophenyl)-5-(4-(morpholine-4-carbonyl)phenylamino)oxazole-4-carboxamide, 2-(2-Chloro-6-fluorophenyl)-5-[[4-(4-morpholinylcarbonyl)phenyl]amino]-4-oxazolecarboxamide, SAR 20347, SAR20347

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18ClFN4O4
CAS Number:
Molecular Weight:
444.84
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

Fc1c(c(ccc1)Cl)c2nc(c([o]2)Nc3ccc(cc3)C(=O)N4CCOCC4)C(=O)N

InChI

1S/C21H18ClFN4O4/c22-14-2-1-3-15(23)16(14)19-26-17(18(24)28)20(31-19)25-13-6-4-12(5-7-13)21(29)27-8-10-30-11-9-27/h1-7,25H,8-11H2,(H2,24,28)

InChI key

HEDPDFHTQKEORT-UHFFFAOYSA-N

Biochem/physiol Actions

Orally active, ATP-competitive, TYK2- and JAK1-selective tyrosine kinase inhibitor in vitro and in vivo.
SAR-20347 is an orally active, ATP-competitive, TYK2- and JAK1-selective tyrosine kinase inhibitor (IC50 = 13 nM/TYK2 & 59 nM/JAK1 vs. 0.88 μM/JAK2 & 1.44 μM/JAK3 with 10 μM ATP) that effectively inhibits cellular TYK2- (IL12-induced pSTAT4 IC50 = 107 nM/CD4+ cells, 126 nM/NK-92) and JAK1- (IL6-induced pSTAT3 IC50 = 345 nM/TF-1, 407 nM/CD14+ cells) mediated signaling with little affinity toward 72 other kinases. SAR-20347 suppresses IL22-induced serum amyloid A (by 44%; 50 mg/kg p.o. 30 min prior to 10 μg IL22 i.p.) and alleviates imiquimod-induced psoriasis-like dermatitis (30 min prior and 5 h post daily 62.5 mg 5% imiquimod cream) in mice in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming-Ming Hu et al.
Cell research, 29(3), 193-205 (2019-01-18)
The mechanisms on metabolic regulation of immune responses are still elusive. We show here that viral infection induces immediate-early NF-κB activation independent of viral nucleic acid-triggered signaling, which triggers a rapid transcriptional induction of bile acid (BA) transporter and rate-limiting
Xia Kang et al.
Cell communication and signaling : CCS, 16(1), 42-42 (2018-07-22)
Chronic muscle injury is characteristics of fatty infiltration and fibrosis. Recently, fibro/adipogenic progenitors (FAPs) were found to be indispensable for muscular regeneration while were also responsible for fibrosis and fatty infiltration in muscle injury. Many myokines have been proven to
Melissa G Works et al.
Journal of immunology (Baltimore, Md. : 1950), 193(7), 3278-3287 (2014-08-27)
Psoriasis is a chronic autoimmune disease affecting the skin and characterized by aberrant keratinocyte proliferation and function. Immune cells infiltrate the skin and release proinflammatory cytokines that play important roles in psoriasis. The Th17 network, including IL-23 and IL-22, has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service