Skip to Content
Merck
All Photos(1)

Key Documents

SAB4501618

Sigma-Aldrich

Anti-KCNA1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

HBK1, HUKI, Potassium voltage-gated channel subfamily A member 1, Voltage-gated potassium channel subunit Kv1.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 56 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNA1(3736)

General description

Anti-KCNA1 Antibody detects endogenous levels of total KCNA1 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human KCNA1.

Immunogen Range: 256-305

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Liu et al.
Channels (Austin, Tex.), 13(1), 321-330 (2019-07-30)
Voltage-gated potassium channel subfamily A member 1 (KCNA1/Kv1.1) is an important component of type A potassium channels, which has been found to be involved in various tumors. This study aimed to identify the role of KCNA1 in cervical cancer and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service