Skip to Content
Merck
All Photos(1)

Key Documents

RA444100-EP

Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges

A-4-44, aerosol-tight, with 4 × 100 mL rectangular buckets

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909

feature

to hold 0.2 mL to 100 mL tubes
aerosol-tight
high-speed swing-bucket rotor
with 4 × 100 mL rectangular buckets

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 022637401 (NA)
Eppendorf® 5804709004 (EU)

parameter

4,400 × g max. RCF

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models 5804/5804R/5810/5810R

Looking for similar products? Visit Product Comparison Guide

General description

A wide selections of rotors to fit the Eppendorf 5804 and 5810 centrifuges.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service