Skip to Content
Merck
All Photos(1)

Key Documents

P5801

Sigma-Aldrich

Peptide YY porcine

≥97% (HPLC), synthetic

Synonym(s):

PYY, Tyr-Pro-Ala-Lys-Pro-Glu-Ala-Pro-Gly-Glu-Asp-Ala-Ser-Pro-Glu-Glu-Leu-Ser-Arg-Tyr-Tyr-Ala-Ser-Leu-Arg-His-Tyr-Leu-Asn-Leu-Val-Thr-Arg-Gln-Arg-Tyr-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C190H288N54O57
CAS Number:
Molecular Weight:
4240.65
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

synthetic

Quality Level

Assay

≥97% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

Gene Information

pig ... PYY(445018)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Tyr-Pro-Ala-Lys-Pro-Glu-Ala-Pro-Gly-Glu-Asp-Ala-Ser-Pro-Glu-Glu-Leu-Ser-Arg-Tyr-Tyr-Ala-Ser-Leu-Arg-His-Tyr-Leu-Asn-Leu-Val-Thr-Arg-Gln-Arg-Tyr-NH2

Biochem/physiol Actions

Gut hormone that inhibits both secretin and cholecystokinin-stimulated pancreatic secretion. Endogenous nonselective agonist at NPY receptors.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Onaga et al.
Domestic animal endocrinology, 74, 106523-106523 (2020-08-17)
The present study aimed to determine the primary sequence of ovine xenin and clarify the mRNA expression and peptide localization of xenin in the gastrointestinal tract in sheep. The colocalization of xenin and glucose-dependent insulinotropic polypeptide was also compared in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service