Skip to Content
Merck
All Photos(7)

Key Documents

HPA026077

Sigma-Aldrich

Anti-GATM antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AT, Anti-Glycine amidinotransferase, mitochondrial, Anti-L-arginine:glycine amidinotransferase, Anti-Transamidinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

LGRTLTGWVQRTFQSTQAATASSRNSCAADDKATEPLPKDCPVSSYNEWDPLEEVIVGRAENACVPPFTIEVKANTYEKYWPFYQKQGGHYFPKDHLKKAVAEIEEMCNILKTEGVTVRRPDPIDWSLKYKTPD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GATM(2628)

General description

Glycine amidinotransferase (GATM) is expressed in the kidneys, liver, brain, lungs, salivary glands and skeletal muscles. The gene encoding it is localized on human chromosome 15. It is also referred to as AGAT (L-arginine:glycine amidinotransferase) or transamidinase.

Immunogen

Glycine amidinotransferase, mitochondrial Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Glycine amidinotransferase (GATM) is involved in the formation of ornithine and guanidinoacetate from arginine and glycine. This is an important step in creatine synthesis. Mutations in the gene encoding GATM have been shown to lead to arginine-glycine amidinotransferase deficiency.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70055

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhanding Cui et al.
Nature communications, 15(1), 4813-4813 (2024-06-07)
Porcine Reproductive and Respiratory Syndrome Virus (PRRSV) poses a major threat to the global swine industry, yet effective prevention and control measures remain elusive. This study unveils Nitazoxanide (NTZ) as a potent inhibitor of PRRSV both in vitro and in
Syed Baharom et al.
American journal of physiology. Endocrinology and metabolism, 313(1), E75-E83 (2017-03-23)
Our aim was to determine the disposition of creatine in ovine pregnancy and whether creatine is transferred across the placenta from mother to fetus. Pregnant ewes received either 1) a continuous intravenous infusion of creatine monohydrate or saline from 122
Caro-Lyne DesRoches et al.
Human mutation, 37(9), 926-932 (2016-05-29)
Arginine-glycine amidinotransferase (GATM) deficiency is an autosomal-recessive disorder caused by pathogenic variants in GATM. Clinical features include intellectual disability, hypotonia, and myopathy. Due to normal neurodevelopment in asymptomatic individuals on creatine monotherapy, GATM deficiency is a good candidate for newborn
Jasmine A Luzum et al.
Cell metabolism, 21(4), 622-627 (2015-04-12)
Statin-induced myopathy (SIM) is the most common reason for discontinuation of statin therapy. A polymorphism affecting the gene encoding glycine amidinotransferase (GATM rs9806699 G > A) was previously associated with reduced risk for SIM. Our objective was to replicate the
Marcus E Kleber et al.
Circulation. Cardiovascular genetics, 6(5), 505-513 (2013-09-21)
Low serum levels of the amino acid derivative, homoarginine, have been associated with increased risk of total and cardiovascular mortality. Homoarginine deficiency may be related to renal and heart diseases, but the pathophysiologic role of homoarginine and the genetic regulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service