EP022363573
Eppendorf® Flex-Tubes Microcentrifuge Tubes
volume 1.5 mL, red, pack of 500 tubes
All Photos(1)
About This Item
UNSPSC Code:
41121703
Recommended Products
material
polypropylene
red
sterility
non-sterile
feature
graduated
Flex-Tube lid
packaging
pack of 500 tubes
manufacturer/tradename
Eppendorf® 022363573 (NA)
technique(s)
PCR: suitable
volume
1.5 mL
General description
The easy-open, easy-close design of the Flex-Tube provides convenient, ergonomic, and reliable one-handed operation. Whether sample preparation, centrifugation or storage – the Flex-Tube fit perfectly into your workflows. This is the successor to the renowned first microcentrifuge tube 3810 from Eppendorf. The idea of manufacturing a 1.5 mL plastic test tube was born out of the necessity to introduce an appropriate container into the automatic clinical analyzers developed by Eppendorf in 1962. The tried and tested design with invaluable features shows perfect sealing qualities, excellent chemical stability and optimal construction materials for resisting the demands of centrifugal forces. As they are easy to open and close securely, The Eppendorf Flex-Tubes off er comfort and dependability for sample preparation, centrifugation and storage. One vessel for all applications in molecular biology, chemistry and bioche mistry – safer than ever. The Eppendorf Flex-Tubes can be centrifuged up to 30,000 x g without any problems.
Application
Eppendorf Flex-Tubes Microcentrifuge Tubes are ideal for preparing samples, storage, or centrifugation
Features and Benefits
- Functionality is guaranteed from -86 C to 100 C
- Simple verifi cation of the pipetted volume by clear graduation
- All variants can be autoclaved for subsequent use in sterile environments (121 C, 20 min.)
- Manufactured without the use of slip agents, plasticizers and biocides
- High resistance to chemicals, mechanical stress and temperature extremes – manufactured from virgin polypropylene
- Tube lid is easy to open and close for ergonomic handling
- Easy labeling on frosted lid and surface on the side
- Flex-Tube lid is easy to open and close for ergonomic operation
- Polypropylene provides high resistance to chemicals, mechanical stress, and temperature extremes
- g-Safe: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
- Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results
- Graduation for visual control of the pipetted volume
- Frosted lid and surface on the side for easy labeling
- Ensured functionality from -86 C to 100 C
- Autoclavable when open (121 C, 20 min)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG