OV56
96020759, human ovary, Epithelial
Synonym(s):
OV-56 Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
product name
OV56, 96020759
biological source
human ovary
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial
products
VEGF
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human ovarian serous carcinoma (ascites) stage IV, poorly differentiated
Cell Line Description
The OV56 cell line was established from ascitic fluid derived from a 46-year-old female patient diagnosed with ovarian cancer. The tumour was characterised as stage IV, poorly differentiated. Immunocytochemical staining of the cells was reported to show strong staining for cytokeratin (CK), weak staining for carcinoembryonic antigen (CEA) and strong staining for human milk fat globulin (HMFG-2). Production of vascular endothelial growth factor (VEGF) has been detected by ELISA. The patient had received sequential treatment with cisplatin and cyclophosphamide, cisplatin and etoposide and subsequently tamoxifen.
DNA Profile
STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 8,9
D16S539: 10,13
D5S818: 11,12
D7S820: 10
THO1: 6,9
TPOX: 9,11
vWA: 15,19
CSF1PO: 11
D13S317: 8,9
D16S539: 10,13
D5S818: 11,12
D7S820: 10
THO1: 6,9
TPOX: 9,11
vWA: 15,19
Culture Medium
DMEM:HAMS F12 (1:1) + 2mM Glutamine + 5% Foetal Bovine Serum (FBS) + 0.5 ug/ml hydrocortisone + 10ug/ml insulin
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service