GS-109-V-34
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
human skin
packaging
tube of 5 μg 90110504-DNA-5UG
pkg of vial of cells 90110504-1VL
growth mode
Adherent
karyotype
Not specified
morphology
Fibroblast
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human Caucasian skin fibroblast Gardner′s syndrome
Cell Line Description
Derived from a 6 year old Caucasian female with Gardner′s syndrome, an autosomal dominant condition which predisposes to carcinoma and multiple polyps of the colon. Normal female karyotype.
Application
Tumourigenicity studies
DNA Profile
STR-PCR Data: Amelogenin: X
CSF1PO: 10,11
D13S317: 8,11
D16S539: 11,12
D5S818: 12
D7S820: 8,12
THO1: 6,9.3
TPOX: 10,12
vWA: 14,16
CSF1PO: 10,11
D13S317: 8,11
D16S539: 11,12
D5S818: 12
D7S820: 8,12
THO1: 6,9.3
TPOX: 10,12
vWA: 14,16
Culture Medium
EMEM (HBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 20% Foetal Bovine Serum (FBS).
Subculture Routine
Split semi-confluent cultures 1:2 i.e. seeding at 4x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C. Monolayer should form in a 25cm2 in 3-4 days.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service