Skip to Content
Merck
All Photos(1)

Key Documents

NCTC 4093

90102530, mouse embryo, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

NCTC 4093, 90102530

biological source

mouse embryo

growth mode

Adherent

karyotype

Modal no. 71-73; hypotetraploid

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse C57B1/KaLw embryo

Cell Line Description

Derived by cloning from a parent line produced from normal mouse embryo tissue (C57B1/KaLw). The cells have spontaneously transformed in vitro. The cells are negative for polyoma virus infection but are susceptible to the virus.

Application

Transformation and virus studies: Polyoma virus

Culture Medium

NCTC 135 100% + 2mM Glutamine.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using mechanical methods to dislodge the cells (e.g. scraping); 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service