Skip to Content
Merck
All Photos(1)

Documents

P388.D1 (Clone 3124)

85011439, mouse lymph, Macrophage

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

P388.D1 (Clone 3124), 85011439

biological source

mouse lymph

growth mode

Semi-adherent

karyotype

Not specified

morphology

Macrophage

products

Interleukin 1 (IL-1), cells synthesise lysozyme

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse lymphoid macrophage

Cell Line Description

Derivative of P388 selected for the high production of IL-1. The cells have a high effect or activity in antibody dependent cell mediated cytoxicity.

Culture Medium

L15 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS). Please note CO2 is not required.

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 37°C. Cells attach lightly to surface and should be tapped sharply or may need scraping to be detached. CO2 is not required.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service