Skip to Content
Merck
All Photos(1)

Key Documents

Mero-14

9100101, human lung, Spindle shaped cells

Synonym(s):

Mero14 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

Mero-14, 09100101

biological source

human lung

growth mode

Adherent

karyotype

Number of chromosomes 72-75

morphology

Spindle shaped cells

products

Cytokeratins 7, 8, 18 and 19. Epithelial membrane antigen (EMA)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

A human mesothelioma cell line derived from a malignant mesothelioma. Asbestos exposure

Cell Line Description

Mero-14 was derived from the pleural effusion from a male patient aged >60 with a history of asbestos exposure. The Y chromosome could not be detected in this cell line by short tandem repeat (STR)-PCR analysis when tested at ECACC. It is a known phenomenon that due to the increased genetic instability of cancer cell lines the Y chromosome can be rearranged or lost resulting in lack of detection. The cell line is identical to the source provided by the depositor based on the STR-PCR analysis.

Application

Investigation of the properties of malignant mesothelioma cells and the development of new diagnostic methods.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10
D13S317: 12
D16S539: 12,13
D5S818: 9
D7S820: 10,11
THO1: 8
TPOX: 9,11
vWA: 15,18

Culture Medium

Hams F10 + 15% FCS + 2mM Glutamine

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Population doubling approx 24hrs

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service