Skip to Content
Merck
All Photos(1)

Key Documents

C1615

SAFC

CHO Feed Bioreactor Supplement

Animal Component-free, solution, sterile-filtered, suitable for cell culture

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

sterility

sterile-filtered

form

solution

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

glucose: no
phenol red: no
L-glutamine: no

shipped in

ambient

storage temp.

2-8°C

Application

Feed concentrate formulated to optimize CHO cell growth and recombinant protein production in fed-batch bioreactor cultures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service