Skip to Content
Merck
All Photos(1)

Documents

AXYT10MLC

Pipette tips

macro tips, size 10 mL, clear, pkg of 10x200tips/cs

Synonym(s):

10 mL Pipette Tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02

manufacturer/tradename

Axygen Inc T-10ML-C
Corning

packaging

pkg of 10x200tips/cs

size

10 mL

color

clear

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service