Skip to Content
Merck
All Photos(5)

Key Documents

AV54486

Sigma-Aldrich

Anti-ACADVL antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ACAD6, Anti-Acyl-coenzyme A dehydrogenase, very long chain, Anti-LCACD, Anti-VLCAD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
WB
clone:
polyclonal
species reactivity:
horse, dog, rat, pig, bovine, human
citations:
3
technique(s):
immunohistochemistry: suitable
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

64 kDa

species reactivity

horse, dog, rat, pig, bovine, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACADVL(37)

Immunogen

Synthetic peptide directed towards the N terminal region of human ACADVL

Application

Anti-ACADVL antibody produced in rabbit is suitable for western blotting at a concentration of 1μg/mL.

Biochem/physiol Actions

The protein encoded by ACADVL (Acyl-coenzyme A dehydrogenase, very long chain) gene belongs to acyl-CoA dehydrogenases (ACADs) family and is specific to esters of long-chain and very long chain fatty acids like-palmitoyl-CoA, mysritoyl-CoA and stearoyl-CoA. It is a homodimer of 67kDa subunits located in mitochondrion inner membrane, where it catalyzes the initial, rate-limiting step of mitochondrial fatty acid β-oxidation. Missense mutations in ACADVL gene may leads to partial carnitine palmitoyltransferase II deficiency. Deficiency of the encoded protein reduces myocardial fatty acid β-oxidation and results in hypertrophic cardiomyopathy.

Sequence

Synthetic peptide located within the following region: RPYAGGAAQESKSFAVGMFKGQLTTDQVFPYPSVLNEEQTQFLKELVEPV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Tucci et al.
Biochimica et biophysica acta, 1842(5), 677-685 (2014-02-18)
Hypertrophic cardiomyopathy is a typical manifestation of very long-chain acyl-CoA dehydrogenase deficiency (VLCADD), the most common long-chain β-oxidation defects in humans; however in some patients cardiac function is fully compensated. Cardiomyopathy may also be reversed by supplementation of medium-chain triglycerides
Paul J Isackson et al.
Muscle & nerve, 47(2), 224-229 (2012-11-22)
Twenty-six patients with clinical symptoms of adult onset carnitine palmitoyltransferase II (CPTII) deficiency were examined. All patients had skeletal muscle CPTII enzyme activity levels indicative of heterozygosity for CPT2 mutations, however sequence analysis identified no pathogenic mutations within the CPT2
Ryan P McAndrew et al.
The Journal of biological chemistry, 283(14), 9435-9443 (2008-01-30)
Very-long-chain acyl-CoA dehydrogenase (VLCAD) is a member of the family of acyl-CoA dehydrogenases (ACADs). Unlike the other ACADs, which are soluble homotetramers, VLCAD is a homodimer associated with the mitochondrial membrane. VLCAD also possesses an additional 180 residues in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service