Skip to Content
Merck
All Photos(1)

Key Documents

A5236

Sigma-Aldrich

Anti-ASF1 antibody, Mouse monoclonal

clone 4A1/3, purified from hybridoma cell culture

Synonym(s):

Anti-ASF1A, Anti-Silencing function 1 homolog A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

4A1/3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~23 kDa

species reactivity

mouse, human

concentration

~1.0 mg/mL

technique(s)

western blot: 2-4 μg/mL using whole extract of human Jurkat or Raji cells

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ASF1A(25842)

General description

ASF1 is a histone chaperone protein that binds H3 and H4. It functions in nucleosome assembly.
Monoclonal Anti-ASF1 (mouse IgG2a isotype) is derived from the hybridoma 4A1/3 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with an ASF1A fusion protein. ASF1 (anti-silencing function 1) encodes a member of the H3/H4 family of histone chaperone proteins and is similar to the anti-silencing function-1 gene in yeast.

Immunogen

ASF1A fusion protein

Application

Monoclonal Anti-ASF1 antibody produced in mouse has been used in immunoblotting.

Biochem/physiol Actions

ASF1 (anti-silencing function 1) protein is a key component of a histone donor complex. It interacts with histones H3 and H4 and functions together with a chromatin assembly factor 1 (CAF-1) during DNA replication and repair. ASF1 also contributes to genome stability during S phase. Cells lacking ASF1 were found to be sensitive to a number of genotoxic agents that predominantly cause DNA damage by interfering with DNA replication fork progression such as hydroxyurea and camptothecin.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Differential requirements for Tousled-like kinases 1 and 2 in mammalian development
Segura-BS, et al.
Cell Death and Differentiation, 24(11), 1872-1872 (2017)
ANTI-SILENCING FUNCTION1 proteins are involved in ultraviolet-induced DNA damage repair and are cell cycle regulated by E2F transcription factors in Arabidopsis
Lario LD, et al.
Plant Physiology, 162(2), 1164-1177 (2013)
Histone deposition protein Asf1 maintains DNA replisome integrity and interacts with replication factor C
Franco AA, et al.
Genes & Development, 19(11), 1365-1375 (2005)
Sandra Segura-Bayona et al.
Cell death and differentiation, 24(11), 1872-1885 (2017-07-15)
The regulation of chromatin structure is critical for a wide range of essential cellular processes. The Tousled-like kinases, TLK1 and TLK2, regulate ASF1, a histone H3/H4 chaperone, and likely other substrates, and their activity has been implicated in transcription, DNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service