Skip to Content
Merck
All Photos(1)

Key Documents

D1000

General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified

viscosity 990.0 mPa.s (25 °C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

viscosity

990.0 mPa.s(25 °C)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

Find detailed technical data for all Paragon General Viscosity Standards on the Paragon Scientific webpage
Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon General Viscosity Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sumit Pramanik et al.
Scientific reports, 5, 9806-9806 (2015-05-08)
Scaffold design from xenogeneic bone has the potential for tissue engineering (TE). However, major difficulties impede this potential, such as the wide range of properties in natural bone. In this study, sintered cortical bones from different parts of a bovine-femur
Marta López-García et al.
Food chemistry, 199, 597-604 (2016-01-19)
In this work MALDI-TOF mass spectroscopy was investigated to characterise the β-glucan profiles of several commercial health supplements, without any derivatisation or purification pre-treatment. The effect of two solvents (water and dimethyl sulfoxide) and two MALDI matrices (2,5-dihydroxybenzoic acid and
B R Achyut et al.
PLoS genetics, 9(2), e1003251-e1003251 (2013-02-15)
Deletion of tumor suppressor genes in stromal fibroblasts induces epithelial cancer development, suggesting an important role of stroma in epithelial homoeostasis. However, the underlying mechanisms remain to be elucidated. Here we report that deletion of the gene encoding TGFβ receptor
Nicolas Floc'h et al.
Molecular cancer therapeutics, 17(5), 885-896 (2018-02-28)
EGFR exon 20 insertions (Ex20Ins) account for 4% to 10% of EGFR activating mutations in non-small cell lung cancer (NSCLC). EGFR Ex20Ins tumors are generally unresponsive to first- and second-generation EGFR inhibitors, and current standard of care for NSCLC patients
Yumiao Zhang et al.
Nature communications, 7, 11649-11649 (2016-05-20)
Injectable hydrophobic drugs are typically dissolved in surfactants and non-aqueous solvents which can induce negative side-effects. Alternatives like 'top-down' fine milling of excipient-free injectable drug suspensions are not yet clinically viable and 'bottom-up' self-assembled delivery systems usually substitute one solubilizing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service