Skip to Content
Merck
All Photos(1)

Key Documents

95580

Supelco

Xanthydrol

for the detection of urea, ≥99.0% (HPLC)

Synonym(s):

9-Hydroxyxanthene, 9-Xanthenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10O2
CAS Number:
Molecular Weight:
198.22
Beilstein:
10395
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥99.0% (HPLC)

form

powder

quality

for the detection of urea

impurities

~1% xanthone

mp

123-128 °C
127-128 °C (lit.)

functional group

hydroxyl

storage temp.

2-8°C

SMILES string

OC1c2ccccc2Oc3ccccc13

InChI

1S/C13H10O2/c14-13-9-5-1-3-7-11(9)15-12-8-4-2-6-10(12)13/h1-8,13-14H

InChI key

JFRMYMMIJXLMBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Xanthydrol may be used as a derivatization agent for the following:
  • Analysis of urea in urine and wine samples using high-performance liquid chromatography (HPLC) technique.
  • Analysis of trace levels of carbamate pesticides in surface water by gas chromatography–mass spectrometry (GC-MS).

Other Notes

Reagent for the determination of urea

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trace level determinations of carbamate pesticides in surface water by gas chromatography?mass spectrometry after derivatization with 9-xanthydrol.
Yang Y-E and Shin S-H
Journal of Chromatography A, 1305(1-2), 328-332 (2013)
R.W. Kiser et al.
Analytical Chemistry, 33, 314-314 (1961)
Shona Clark et al.
Journal of chromatography. A, 1161(1-2), 207-213 (2007-06-16)
A high-performance liquid chromatography (HPLC) method for the determination of urea that incorporates automated derivatisation with xanthydrol (9H-xanthen-9-ol) is described. Unlike the classic xanthydrol approach for the determination of urea, which involves the precipitation of dixanthylurea (N,N'-di-9H-xanthen-9-ylurea), the derivatisation procedure
P V Biles et al.
Journal of AOAC International, 81(6), 1155-1161 (1998-12-16)
Contamination of food and food packaging material by rodent urine is evidence of insanitary conditions. Urea from rodent urine is used as a chemical indicator of contamination. The limit of detection of the xanthydrol/urea AOAC Method 959.14 by formation of
Kumiko Yamazaki et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 29(5), 705-715 (2012-01-20)
A novel GC-MS method was developed for the determination of acrylamide, which is applicable to a variety of processed foods, including potato snacks, corn snacks, biscuits, instant noodles, coffee, soy sauces and miso (fermented soy bean paste). The method involves

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service