Skip to Content
Merck
All Photos(2)

Key Documents

64012BC

Millipore

MVP ICON® ATP Calibrator Set

MVP ICON®, ATP Luminometer Calibration

Synonym(s):

ATP Calibrator Set, ATP Measurement Calibration Kit, Quality Control ATP Calibrator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NB.79

Agency

EP
ISO
USP

product line

MVP ICON®

shelf life

5 yr

feature

ready-to-use

manufacturer/tradename

MVP ICON®

packaging

set of calibrator rods

storage condition

protect from light (Keep in a tightlyclosed container, away from any light sources)

application(s)

food and beverages

compatibility

for use with MVP ICON instrument

storage temp.

15-25°C

General description

MVP ICON® ATP Calibrator Set consists of two calibration devices (“rods”) that are used for performance verification and calibration of the MVP ICON® and MVP systems. The luminescent calibrator consists of a 14C radioactive source that emits a very low level (5 x 105 d.p.m., or 0.225 microcuries) of low-energy ß-radiation in a plastic scintillation matrix. This matrix is configured to give constant light output, within 10 % of its original value, at a sufficiently high level to make consistent verification and calibration functions possible over the five‐year life of the calibrator. The non‐luminescent black plastic calibrator provides a reliable low‐end or “dark” source reading.

Application

MVP ICON® ATP Calibrator Set is used for performance verification and calibration of the MVP ICON® instrument for rapid monitoring of surface hygiene in the Food and Beverage industries.

Features and Benefits

  • On-site calibration
  • Satisfies HACCP and GMP requirements
  • No need to send the equipment out to a 3rd party.

Components

  • One luminescent calibrator
  • One non-luminescent calibrator

Suitability

ATP Testing, Food Processing, HACCP Monitoring, Sanitation

Principle

The luminescent calibrator consists of a 14C radioactive source that emits a very low level (5 x 105 d.p.m., or 0.225 microcuries) of low energy ß radiation in a plastic scintillation matrix. This matrix is configured to give constant light output, within 10% of its original value, at a sufficiently high level to make consistent verification and calibration functions possible over the five- year life of the calibrator. The non-luminescent black plastic calibrator provides a reliable low-end or “dark” source reading.

Testing Method

ATP Bioluminescence

Storage and Stability

To ensure the stability and longevity of the luminescent calibrator, the following precautions must be taken:
  • Limit Exposure to Light.
  • Store the luminescent calibrator in the dark or weak ambient light.
  • If the luminescent calibrator is exposed to bright light before use, insert it in the MVP ICON® or MVP and wait a minimum of 2 minutes before proceeding.
  • The kit box provided is recommended as an appropriate dark storage.
  • Do not expose to direct sunlight or intense artificial light before use.

Other Notes

Avoid crushing, cutting, or dropping the calibrators.

Legal Information

BioControl is a registered trademark of BioControl Systems, Inc.
MVP ICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Kit Components Also Available Separately

Product No.
Description
SDS

  • 72001SBCLuminescent Calibrator RodSDS

  • Non Luminescent Calibrator Rod

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service