49161
D-(+)-Glucose monohydrate
tested according to Ph. Eur.
Synonym(s):
Dextrosum (Glucosum) monohydricum, Dextrose monohydrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H12O6 · H2O
CAS Number:
Molecular Weight:
198.17
Beilstein:
5250278
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21
Recommended Products
Agency
USP/NF
tested according to Ph. Eur.
Quality Level
Assay
97.5-102.0% anhydrous basis (HPLC)
form
powder
color
white
SMILES string
O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O
InChI
1S/C6H12O6.H2O/c7-1-3(9)5(11)6(12)4(10)2-8;/h1,3-6,8-12H,2H2;1H2/t3-,4+,5+,6+;/m0./s1
InChI key
SPFMQWBKVUQXJV-BTVCFUMJSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Effects of mammalian blood with different glucose levels on reproduction, growth and survival of the southern medicinal leech, Hirudo verbana Carena, 1820.: This study explores the effects of varying glucose levels in mammalian blood on the reproduction, growth, and survival of the medicinal leech. The research highlights the role of D-(+)-Glucose monohydrate in biological systems and its potential applications in aquaculture and biomedical research. (Karataş et al., 2022).
- Evaluation of the variations in chemical and microbiological properties of the sourdoughs produced with selected lactic acid bacteria strains during fermentation.: This research evaluates the chemical and microbiological changes in sourdoughs fermented with specific lactic acid bacteria, with a focus on the role of D-(+)-Glucose monohydrate in the fermentation process. The findings are significant for the food industry, particularly in the production of high-quality sourdough bread. (Boyaci Gunduz et al., 2022).
Other Notes
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Katarina Akhmetova et al.
eLife, 10 (2021-09-02)
Stimulator of interferon genes (STING) plays an important role in innate immunity by controlling type I interferon response against invaded pathogens. In this work, we describe a previously unknown role of STING in lipid metabolism in Drosophila. Flies with STING
Daphne H E W Huberts et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(32), 11727-11731 (2014-07-30)
Calorie restriction (CR) is often described as the most robust manner to extend lifespan in a large variety of organisms. Hence, considerable research effort is directed toward understanding the mechanisms underlying CR, especially in the yeast Saccharomyces cerevisiae. However, the
Sweta Rathore et al.
Journal of microencapsulation, 31(5), 469-478 (2014-04-05)
This work evaluated the feasibility of microencapsulating Clostridium acetobutylicum ATCC 824 cells by emulsification for fermentation to produce biobutanol. The effects of selected emulsification process on viability of the vegetative cells and spores were investigated to enable the selection of
Jesper T Andreasen et al.
Brain research, 1601, 117-126 (2015-01-13)
Depression and anxiety often co-occur, and conventional monoamine-facilitating antidepressants show efficacy against symptoms in both disorders. Rodent studies indicate that antidepressant effects of monoamine-based antidepressants involve increased α-amino-3-hydroxy-5- methyl-4-isoxazolepropionic acid glutamate receptor (AMPAR) neurotransmission, and positive allosteric modulators (PAMs) at
Brooke Tata et al.
PLoS biology, 12(9), e1001952-e1001952 (2014-09-24)
Characterization of the genetic defects causing gonadotropic deficiency has made a major contribution to elucidation of the fundamental role of Kisspeptins and Neurokinin B in puberty onset and reproduction. The absence of puberty may also reveal neurodevelopmental disorders caused by
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service