Skip to Content
Merck
All Photos(1)

Documents

34079

Supelco

Carfentrazone-ethyl

PESTANAL®, analytical standard

Synonym(s):

Ethyl 2-chloro-3-{2-chloro-5-[4-(difluoromethyl)-4,5-dihydro-3-methyl-5-oxo-1H-1,2,4-triazol-1-yl]-4-fluoro-phenyl}propionate, Ethyl-α,2-dichloro-5-[4-(difluoromethyl)-4,5-dihydro-3-methyl-5-oxo-1H-1,2,4-triazol-1-yl]-4-fluorobenzenepropanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14Cl2F3N3O3
CAS Number:
Molecular Weight:
412.19
Beilstein:
9081231
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOC(=O)C(Cl)Cc1cc(N2N=C(C)N(C(F)F)C2=O)c(F)cc1Cl

InChI

1S/C15H14Cl2F3N3O3/c1-3-26-13(24)10(17)4-8-5-12(11(18)6-9(8)16)23-15(25)22(14(19)20)7(2)21-23/h5-6,10,14H,3-4H2,1-2H3

InChI key

MLKCGVHIFJBRCD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Carfentrazone-ethyl is an aryl triazolinone reduced-risk herbicide used in the cultivation of wheat, soybean and corn.

Application

Carfentrazone-ethyl may be used as a reference standard in the determination of carfentrazone-ethyl in:
  • Water samples using direct immersion solid-phase microextraction (DI-SPME) with gas chromatography coupled with mass spectrometry (GC-MS).
  • Honey samples using liquid chromatography-electrospray ionization-tandem mass spectrometry (LC-ESI-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Degradation of [14C] carfentrazone-ethyl under aerobic aquatic conditions.
Elmarakby A S, et al.
Journal of Agricultural and Food Chemistry, 49(11), 5285-5293 (2001)
Levels of 13 multi-class pesticide residues in Polish honeys determined by LC-ESI-MS/MS.
Kujawski W M and Namiesnik J.
Food Control, 22(6), 914-919 (2011)
Multi-residue method for the determination of 16 recently used pesticides from various chemical groups in aqueous samples by using DI-SPME coupled with GC--MS
Tankiewicz M, et al.
Talanta, 107, 1-10 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service