Skip to Content
Merck
All Photos(1)

Key Documents

23128

Supelco

L-Lysine

analytical standard

Synonym(s):

(S)-2,6-Diaminocaproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)4CH(NH2)CO2H
CAS Number:
Molecular Weight:
146.19
Beilstein:
1722531
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

analyte chemical class(es)

amino acids, peptides, proteins

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤2.0% water

color

white to off-white

mp

215 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

NCCCC[C@H](N)C(O)=O

InChI

1S/C6H14N2O2/c7-4-2-1-3-5(8)6(9)10/h5H,1-4,7-8H2,(H,9,10)/t5-/m0/s1

InChI key

KDXKERNSBIXSRK-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Lysine is an essential amino acid which is required for human and animal growth. It belongs to the aspartate family of amino acids.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Engineering photosynthetic production of L-lysine.
Korosh, T. C
Metabolic engineering, 44, 273-283 (2017)
J Robin Highley et al.
Neuropathology and applied neurobiology, 40(6), 670-685 (2014-04-23)
Loss of nuclear TDP-43 characterizes sporadic and most familial forms of amyotrophic lateral sclerosis (ALS). TDP-43 (encoded by TARDBP) has multiple roles in RNA processing. We aimed to determine whether (1) RNA splicing dysregulation is present in lower motor neurones
Nicholas I Proellocks et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(7), 3103-3113 (2014-04-08)
The genomes of malaria parasites (Plasmodium spp.) contain a family of genes encoding proteins with a Plasmodium helical interspersed subtelomeric (PHIST) domain, most of which are predicted to be exported into the parasite-infected human red blood cell (iRBC). Here, using
Audrey Letourneau et al.
Nature, 508(7496), 345-350 (2014-04-18)
Trisomy 21 is the most frequent genetic cause of cognitive impairment. To assess the perturbations of gene expression in trisomy 21, and to eliminate the noise of genomic variability, we studied the transcriptome of fetal fibroblasts from a pair of
Hillary F Allen et al.
The Biochemical journal, 459(3), 505-512 (2014-03-01)
The tandem PHD (plant homeodomain) fingers of the CHD4 (chromodomain helicase DNA-binding protein 4) ATPase are epigenetic readers that bind either unmodified histone H3 tails or H3K9me3 (histone H3 trimethylated at Lys⁹). This dual function is necessary for the transcriptional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service