Skip to Content
Merck
All Photos(1)

Key Documents

07424

Sigma-Aldrich

1-Ethyl-3-methylimidazolium thiocyanate

for electrochemistry, ≥99.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11N3S
CAS Number:
Molecular Weight:
169.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (T)

impurities

≤0.2% water

refractive index

n20/D 1.553

conductivity

~20.2 mS/cm

mp

-6 °C

anion traces

sulfate (SO42-): ≤500 mg/kg

SMILES string

[S-]C#N.CCn1cc[n+](C)c1

InChI

1S/C6H11N2.CHNS/c1-3-8-5-4-7(2)6-8;2-1-3/h4-6H,3H2,1-2H3;3H/q+1;/p-1

InChI key

VASPYXGQVWPGAB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Ionic Liquids serve as non-aqueous electrolytes in electrochemical applications, valued for their conductivity and stability.

Ionic Liquids serve as non-aqueous electrolytes in electrochemical applications, valued for their conductivity and stability.

Ionic Liquids serve as non-aqueous electrolytes in electrochemical applications, valued for their conductivity and stability.

Ionic Liquids serve as non-aqueous electrolytes in electrochemical applications, valued for their conductivity and stability.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service