Skip to Content
Merck
All Photos(1)

Documents

00933

Sigma-Aldrich

4-Acetylbenzenesulfonyl chloride

≥95.0% (AT)

Synonym(s):

Acetophenone-4-sulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7ClO3S
CAS Number:
Molecular Weight:
218.66
Beilstein:
2099148
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (AT)

form

powder

mp

84-87 °C

SMILES string

CC(=O)c1ccc(cc1)S(Cl)(=O)=O

InChI

1S/C8H7ClO3S/c1-6(10)7-2-4-8(5-3-7)13(9,11)12/h2-5H,1H3

InChI key

FXVDNCRTKXMSEZ-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service