Skip to Content
Merck
All Photos(1)

Key Documents

03-1660

Sigma-Aldrich

Benzyl alcohol

SAJ special grade, ≥99.0%

Synonym(s):

Benzenemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
878307
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

grade

SAJ special grade

vapor density

3.7 (vs air)

vapor pressure

13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

817 °F

expl. lim.

0.34-6.3 % (lit.)

availability

available only in Japan

refractive index

n20/D 1.539 (lit.)

bp

203-205 °C (lit.)

mp

−16-−13 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

SMILES string

OCc1ccccc1

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

WVDDGKGOMKODPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

213.8 °F - DIN 51758

Flash Point(C)

101 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C L Froebe et al.
Dermatologica, 181(4), 277-283 (1990-01-01)
The relationship between the in vivo irritation potential of sodium lauryl sulfate (SLS) and linear alkyl benzene sulfonate (LAS) and the ability of these two surfactants to remove lipid from the stratum corneum (SC) in vitro were investigated. Either surfactant
Béatrice Hechler et al.
The Journal of pharmacology and experimental therapeutics, 314(1), 232-243 (2005-03-29)
Our aim was to determine whether the newly described P2X1 antagonist NF449 [4,4',4'',4'''-(carbonylbis(imino-5,1,3-benzenetriylbis(carbonylimino)))tetrakis-benzene-1,3-disulfonic acid octasodium salt] could selectively antagonize the platelet P2X1 receptor and how it affected platelet function. NF449 inhibited alpha,beta-methyleneadenosine 5'-triphosphate-induced shape change (IC50 = 83 +/- 13
D J Greenblatt et al.
Arzneimittel-Forschung, 33(12), 1671-1673 (1983-01-01)
The non-steroidal antiinflammatory agent ibuprofen (Motrin) can be reliably quantitated in body fluids using high-pressure liquid chromatography with a newly developed radial compression separation system. After addition of the structural analogue flurbiprofen as internal standard, biological samples (plasma or cerebrospinal
Kelly Roberta Francisco et al.
Langmuir : the ACS journal of surfaces and colloids, 28(41), 14531-14539 (2012-09-13)
Polymers formed by the self-assembly of a bis(urea)-based polymer, 2,4-bis(2-ethylhexylureido)toluene (EHUT), in organic solvents such as octane are promising systems with remarkable rheological properties. This is the first self-assembled polymer recently reported as a hydrodynamic drag reducer for hydrocarbons. The
H I Maibach et al.
Archives of environmental health, 36(5), 256-260 (1981-09-01)
Penetration of benzene through the skin of the rhesus monkey was determined using 14C-benzene, and quantitating the labelled metabolites in urine. The modes of application and amounts of benzene that penetrated the skin (indicated in parentheses) are as follows: (1)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service