Skip to Content
Merck
All Photos(1)

Documents

SCC190

Sigma-Aldrich

J14 Human SLP-76-Deficient Jurkat Cell Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.45

description

Cell line type: Blood Cell

technique(s)

cell culture | mammalian: suitable

General description

J14 cells are a unique Jurkat-derived human T cell line that lacks expression of SLP-76 (SH2 domain–containing leukocyte protein of 76 kilodaltons), an adaptor protein and PTK substrate expressed in hematopoietic cells (2). SLP-76 together with the scaffolding protein LAT act as a hub for various signaling effectors (1). Except SLP-76, the expression profile of J14 cells encompasses all necessary components of downstream pathways including those required for IL-2 cytokine induction (3), making the J14 cell line ideal for mutational analysis and introduction of reporter constructs.

Source:
J14 cells are a clonal derivative of Jurkat, a T cell line derived from bone marrow of a 14-year old male (4). Jurkat cells were transfected with an episomal vector and selected for hygromycin resistance. The transgene was subsequently lost via non-selective growth (2).
T-cell-receptor signaling has a critical role in adaptive immune responses and has been extensively studied as a general model for complex cellular responses (1). T-cell-receptor signaling to downstream pathways is mediated by non-receptor protein tyrosine kinases (PTKs), which interact with substrate proteins in a complex network.

References:
1. J Cell Sci. 2009; 122(Pt 9): 1269-1273.
2. J Biol Chem. 1995; 270(13): 7029-7032.
3. Science. 1998; 281(5375): 413-416.
4. Int J Cancer. 1977; 19(5): 621-626.

Application

This product is sold solely for research use per the terms of the “Restricted Use Agreement” which govern its use as detailed in the product Data Sheet or Certificate of Analysis. For information regarding any other use, please contact licensing@emdmillipore.com.

Quality

  • Each vial contains >=1X106 viable cells.
  • Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.
  • Cells are verified to be of human origin and negative for inter-species contamination from rat, mouse, chinese hamster, Golden Syrian hamster, and non-human primate (NHP) as assessed by a Contamination CLEAR panel by Charles River Animal Diagnostic Services.
  • Cells are negative for mycoplasma contamination.
  • Each lot of cells is genotyped by STR analysis to verify the unique identity of the cell line.

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service