PX1480
Potassium hydroxide
pellets, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS
Synonym(s):
Caustic Potash, Caustic potash
About This Item
Recommended Products
vapor pressure
1 mmHg ( 719 °C)
Quality Level
Assay
≥85.0%
form
pellets
color
white to off-white
pH
14 (20 °C, 56 g/L in H2O)
mp
361 °C (lit.)
solubility
1130 g/L
anion traces
carbonate (K2CO3): ≤2.0%
chloride (Cl-): ≤0.01%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.003%
cation traces
Ca: ≤0.005%
Fe: ≤0.001%
Mg: ≤0.002%
N: ≤0.001%
Na: ≤0.05%
Ni: ≤0.001%
heavy metals: ≤0.001%
shipped in
ambient
storage temp.
room temp
SMILES string
[OH-].[K+]
InChI
1S/K.H2O/h;1H2/q+1;/p-1
InChI key
KWYUFKZDYYNOTN-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Analysis Note
Calcium (Ca): 0.005% max
Chloride (Cl): 0.01% max
Color: White to off-white
Form: Pellets
Heavy Metals (by ICP-OES): 0.001% max
Iron (Fe): 0.001% max
Magnesium (Mg): 0.002% max
Nickel (Ni): 0.001% max
Nitrogen compounds (as N): 0.001% max
Phosphate (PO4): 5ppm max
Potassium carbonate (K2CO3): 2.0% max
Sodium (Na): 0.05% max
Sulfate (SO4): 0.003% max
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service