Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB337F

Sigma-Aldrich

Milli-Mark® Anti-Myelin Associated Glycoprotein-FITC Antibody, clone 513

clone 513, Milli-Mark®, from mouse

Synonym(s):

Brain neuron cytoplasmic protein 3, Sialic acid-binding Ig-like lectin 4a, Siglec-4a, MAG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

513, monoclonal

species reactivity

rat

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

rat ... Mag(29409)

General description

Myelin-associated glycoprotein (MAG), a member of the immunoglobulin superfamily, is thought to be involved in the process of myelination. Axonal regeneration in the adult central nervous system (CNS) is limited by two proteins in myelin; Nogo and MAG. MAG has been implicated in the inhibition of nerve regeneration in the CNS.

Specificity

Antibody recognizes Myelin Associated Glycoprotein.

Immunogen

L2 epitope carrying glycoprotein extract of 1-2 old day chicken brain.

Application

Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers
This Milli-Mark Anti-Myelin Associated Glycoprotein-FITC Antibody, clone 513 is validated for use in FC for the detection of Myelin Associated Glycoprotein.

Quality

Evaluated by flow cytometry using B35 cells

Target description

69.3 kDa Calculated

Physical form

Protein A purified
Purified mouse monoclonal IgG1 conjugated to FITC in PBS with 0.1% sodium azide and 15 mg/mL BSA

Storage and Stability

Maintain refrigerated at 2-8°C protected from light for up to 6 months from date of receipt.

Analysis Note

Control
B35 cells

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service