Skip to Content
Merck
All Photos(1)

Key Documents

AB16527

Sigma-Aldrich

Anti-MyD88 Antibody, CT

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MYD88(4615)

Specificity

The pro-inflammatory cytokine IL-1 induced cellular response requires IL-1 receptor complex including IL-1RI and IL-RAcP. Recently, MyD88 was identified as an adapter molecule in the IL-1 signaling pathway (Muzio et al. 1997). MyD88 associates with and recruits IRAK to the IL-1 receptor complex in response to IL-1 treatment and dominant negative form of MyD88 attenuates IL-1R mediated NF-kB activation. MyD88 is also employed as a regulator molecule by IL-18 receptor and human Toll receptor (Adachi et al. 1998; Medzhitov et al. 1998), which are members in the Toll/IL-1R family of receptors. Targeted disruption of the MyD88 gene results in loss of cellular responses to IL-1 and IL-18, and MyD88-defecient mice lack responses to bacterial product LPS that employs Toll-like receptors 2 and 4 (TLR2 and TLR4) as the signaling receptors. MyD88 is a general adapter protein for the Toll/1R family of receptors and plays an important role in the inflammatory response induced by cytokines IL-1 and IL-18 and endotoxin. MyD88 gene is expressed in many tissues.

Immunogen

Epitope: C-terminus
Rabbit anti-MyD88 (C-terminus) polyclonal antibody was raised against a peptide corresponding to amino acids 279 to 296 of human MyD88 (Bonnert et al. 1997), which are identical to those of mouse.

Application

Anti-MyD88 Antibody, C-terminus is an antibody against MyD88 for use in WB.
Research Category
Inflammation & Immunology
Research Sub Category
Inflammation & Autoimmune Mechanisms
Western blot: 1:500 to 1:1000

Whole cell lysate from Jurkat cells can be used as positive control and a 35 kDa band should be detected.

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in PBS containing 0.02% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naturally occurring mutation affecting the MyD88-binding site of TNFRSF13B impairs triggering of class switch recombination.
Almejun, MB; Cols, M; Zelazko, M; Oleastro, M; Cerutti, A; Oppezzo, P; Cunningham-Rundles et al.
European Journal of Immunology null
Ramesh K Paidi et al.
Journal of immunology (Baltimore, Md. : 1950), 207(10), 2521-2533 (2021-10-15)
Many patients with coronavirus disease 2019 in intensive care units suffer from cytokine storm. Although anti-inflammatory therapies are available to treat the problem, very often, these treatments cause immunosuppression. Because angiotensin-converting enzyme 2 (ACE2) on host cells serves as the
The transmembrane activator TACI triggers immunoglobulin class switching by activating B cells through the adaptor MyD88.
He, B; Santamaria, R; Xu, W; Cols, M; Chen, K; Puga, I; Shan, M; Xiong, H; Bussel et al.
Nature Immunology null
Kendra A Bussey et al.
Journal of virology, 88(16), 9245-9259 (2014-06-06)
The human pathogen Kaposi's sarcoma-associated herpesvirus (KSHV), the etiological agent of Kaposi's sarcoma, primary effusion lymphoma, and multicentric Castleman's disease, establishes lifelong latency upon infection. Murine gammaherpesvirus 68 (MHV68) is a well-established model for KSHV. Toll-like receptors (TLRs) play a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service